Human Recombinant ATG13 (from E. coli)
Supplier: Rockland Immunochemicals
009-001-R44S 009-001-R44
10800-194EA 364.21 USD
10800-194 10800-196
Human Recombinant ATG13 (from E. coli)
Proteins and Peptides
ATG13 is a protein that is part of the autophagy complex which is the major route by which cytoplasmic contents are delivered to the lysosome for degradation. ATG13 localizes on the autophagic isolation membrane and is essential for autophagosome formation. In mammals, ATG13 interacts with ATG1 (ULK1/2) and FIP200 to form the autophagy. ATG101 can also bind to ATG13 protein and is important for the stability and basal phosphorylation of ATG13 and ULK1 (1). mTOR can suppress the autophagy process through direct regulation of the ULK1-Atg13-FIP200 complex where mTOR has been shown to phosphorylate ATG13 (2). ATG13 Protein is ideal for investigators involved in Signaling Proteins, Apoptosis Proteins, Angiogenesis, Cardiovascular Disease, Cellular Stress, Inflammation, Metabolic Disorder, Neurobiology, NfkB Pathway, and p38 Pathway research.