Human Recombinant ATF2 (from E. coli)
Supplier: Rockland Immunochemicals
009-001-R43S 009-001-R43
10800-158EA 364.21 USD
10800-158 10801-950
Human Recombinant ATF2 (from E. coli)
Proteins and Peptides
Activating transcription factor (ATF)-2, also designated CRE-BP1, is a member of the ATF/cyclic AMP-responsive element binding protein family of transcription factors. ATF2 has two unique characteristics: it mediates the adenovirus E1A-induced trans-activation and forms a heterodimer with c-Jun (1). ATF2 is a target of the JNK signal transduction pathway (2). ATF2 is phosphorylated by JNK on two closely spaced threonine residues within the NH2-terminal activation domain. The replacement of these phosphorylation sites with alanine inhibited the transcriptional activity of ATF2. Furthermore, p38 MAPK can also phosphorylate ATF2. ATF2 Protein is ideal for investigators involved in Signaling Reagents, Protein Substrates, Angiogenesis, Apoptosis/Autophagy, Cardiovascular Disease, Cellular Stress, Inflammation, Metabolic Disorder, Neurobiology, NfkB Pathway, and p38 Pathway research.