Would you like to log in again?
ARAF protein-GST fusion, Rockland Immunochemicals
Supplier: Rockland Immunochemical
009-001-R41S 009-001-R41
10801-894 10801-896
ARAF protein-GST fusion, Rockland Immunochemicals
ARAF belongs to the RAF subfamily of the Ser/Thr protein kinase family, and maybe involved in cell growth and development. ARAF play a critical role in cell growth and development (1).The N-terminal regulatory domain of ARAF interacted with the putative mitochondrial proteins TOM and TIM44. ARAF is expressed predominantly in urogenital tissues (2). The complete coding sequence of the human A-raf-1 oncogene and transforming activity of a human A-raf carrying retrovirus. ARAF Protein is ideal for investigators involved in Signaling Proteins, Cellular Proteins, and Cancer research.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty