Human Recombinant ACVR1C (from Baculovirus (Sf9 Insect cells))
Supplier: Rockland Immunochemicals
009-001-R39S 009-001-R39
10799-962EA 713.69 USD
10799-962 10799-960
Human Recombinant ACVR1C (from Baculovirus (Sf9 Insect cells))
Proteins and Peptides
ALK7 is a type I receptor for the TGFB family of signaling molecules in which type I receptors phosphorylate cytoplasmic SMAD transcription factors, which then translocate to the nucleus and interact directly with DNA or in complex with other transcription factors (1). Alk7 was expressed in all insulin, glucagon, and somatostatin -positive cells of the pancreas (2). GDF3 regulates adipose tissue homeostasis and energy balance under nutrient overload, in part, by signaling through ALK7 and Alk7 showed reduced fat accumulation and partial resistance to diet-induced obesity, similar to Gdf3. ALK7 Protein is ideal for investigators involved in Signaling Proteins, Cellular Proteins, AKT/PKB Pathway, Angiogenesis, Cancer, Cardiovascular Disease, JAK/STAT Pathway, Neurobiology, and Ser/Thr Kinase research.