Would you like to log in again?
14-3-3 sigma protein, Rockland Immunochemicals
Supplier: Rockland Immunochemical
009-001-R32 009-001-R32S
10799-464EA 472.72 USD InStock
10799-464 10799-462
14-3-3 sigma protein, Rockland Immunochemicals
Proteins and Peptides
14-3-3σ or stratifin is a protein that is strongly induced by gamma irradiation and other DNA-damaging agents (1). The induction of 14-3-3σ is mediated by a p53 responsive element. Exogenous introduction of 14-3-3σ into cycling cells results in a G2 cell cycle arrest (2). Knockout of 14-3-3σ in cells showed that the cells are unable to maintain cell cycle arrest after DNA damage. The 14-3-3σ -/- cells die as they enter mitosis. This process is associated with a failure of the 14-3-3σ-deficient cells to sequester the proteins that initiate mitosis and prevent them from entering the nucleus. Thus, 14-3-3σ plays an important role in maintaining the G2 checkpoint in cells and preventing mitotic death. 14-3-3σ Protein is ideal for investigators involved in Cell Stress& Chaperone Proteins, AKT/PKB Pathway, Cancer, Cell Cycle, Cellular Stress, ERK/MAPK Pathway, Neurobiology, PKA/PKC Pathway, and WNT Signaling research.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty