Would you like to log in again?
14-3-3 zeta protein, Rockland Immunochemicals
Supplier: Rockland Immunochemical
009-001-R28S 009-001-R28
10799-478 10799-480
14-3-3 zeta protein, Rockland Immunochemicals
14-3-3ζ (also known as tyrosine 3-monooxygenase/ tryptophan 5-monooxygenase activation protein, zeta polypeptide) is a member of the 14-3-3 family of proteins which mediate signal transduction by binding to phosphoserine-containing proteins. 14-3-3ζ protein plays a key role in cancer biology by being an important regulator of major cellular processes such as proliferation, differentiation, senescence and apoptosis (1). 14-3-3ζ protein has been shown to interact with the IRS1 protein, suggesting a role for this protein in regulating insulin sensitivity by interrupting the association between the insulin receptor and IRS1 (2). 14-3-3ζ Protein is ideal for investigators involved in Cell Stress& Chaperone Proteins, Cell Signaling, AKT/PKB Pathway, Cancer, Cell Cycle, Cellular Stress, ERK/MAPK Pathway, Neurobiology, PKA/PKC Pathway, WNT Signaling research.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty