Human Recombinant AKT2 (from Baculovirus (Insect cells))
Supplier: Rockland Immunochemicals
009-001-P23
10799-944EA 353.78 USD
10799-944
Human Recombinant AKT2 (from Baculovirus (Insect cells))
Proteins and Peptides
AKT2 is a component of the PI-3 kinase pathway and is activated by phosphorylation at Ser 473 and Thr 308. AKT is a cytoplasmic protein also known as Protein Kinase B (PKB) and RAC (Related to A and C kinases). AKT is a key regulator of many signal transduction pathways, and it exhibits tight control over cell proliferation and cell viability. Overexpression or inappropriate activation of AKT is noted in many types of cancer. AKT mediates many of the downstream events of PI 3-kinase (a lipid kinase activated by growth factors, cytokines and insulin). PI 3-kinase recruits AKT to the membrane, where it is activated by PDK1 phosphorylation. Once phosphorylated, AKT dissociates from the membrane and phosphorylates targets in the cytoplasm and the cell nucleus. AKT has two main roles: (i) inhibition of apoptosis; (ii) promotion of proliferation. AKT2 recombinant protein is ideal for investigators involved in Cell Signaling, Neuroscience and Signal Transduction research.