Would you like to log in again?
ERK2 hu recomb protein, Rockland Immunochemicals
Supplier: Rockland Immunochemical
009-001-GQ9
76221-872EA 337.65 USD InStock
76221-872
ERK2 hu recomb protein, Rockland Immunochemicals
Proteins and Peptides
ERK2 recombinant protein was produced in Sf9 cells. Mitogen activiated protein kinase 1, also known as MAPK1, ERK, or ERK2, is an integral component of the MAP kinase cascade that regulates cell growth and differentiation. ERK1 and ERK2 are activated by MEK1 and MEK2 in the B-raf signaling pathway resulting in its translocation to the nucleus where it phosphorylates nuclear targets. Human ERK1 and ERK2 are 84% identical in sequence and share common functionality in cells. Recombinant ERK2 protein is ideal for investigators involved in Neuroscience, Cell Signaling and Cancer Research.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty