Would you like to log in again?
ERK1 hu recomb protein, Rockland Immunochemicals
Supplier: Rockland Immunochemical
009-001-GQ8
76221-870EA 337.65 USD InStock
76221-870
ERK1 hu recomb protein, Rockland Immunochemicals
Proteins and Peptides
ERK1 recombinant protein was produced in Sf9 cells. Mitogen activiated protein kinase 3, also known as MAPK3, ERK, or ERK1, is an integral component of the MAP kinase cascade that regulates cell growth and differentiation. ERK1 and ERK2 are activated by MEK1 and MEK2 in the B-raf signaling pathway resulting in its translocation to the nucleus where it phosphorylates nuclear targets. Human ERK1 and ERK2 are 84% identical in sequence and share common functionality in cells. Recombinant ERK1 protein is ideal for investigators involved in Neuroscience, Cell Signaling and Cancer Research.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty