Human Recombinant IL9 (from E. coli)
Supplier: Rockland Immunochemicals
009-001-B96
RL009-001-B96EA 326.07 USD
RL009-001-B96
Human Recombinant IL9 (from E. coli)
Proteins and Peptides
Interleukin (IL-9) is known to regulate many cell types involved in T- helper type 2 responses classically associated with asthma, including B- and T- lymphocytes, mast cells, eosinophils and epithelial cells. Growing evidence obtained from human genomic analysis and antigen-challenged transgenic mice suggests that IL-9 is a candidate factor in immunoglobulin E (IgE) production and thus is thought to be associated with bronchial inflammation and bronchial hyperresponsiveness (BHR). Recombinant Human IL-9 produced in E.coli is a single, non-glycosylated single polypeptide chain containing 127 amino acids and having a molecular mass of 14,004 Daltons.