Would you like to log in again?
IL-7 Human Recombinant Protein, Rockland Immunochemicals
Supplier: Rockland Immunochemical
009-001-B94
RL009-001-B94
IL-7 Human Recombinant Protein, Rockland Immunochemicals
Interleukin 7 (IL-7), which is constitutively produced particularly by stromal cells from the bone marrow and thymus, plays a crucial role in T cell homeostasis. This cytokine is implicated in thymopoiesis because it sustains thymocyte proliferation and survival. It regulates peripheral naive T cell survival by modulating the expression of the anti-apoptotic molecule Bcl-2, and sustains peripheral T cell expansion in response to antigenic stimulation. Infection by the human immunodeficiency virus (HIV) leads to severe T lymphopenia and general immune dysfunction. Increased IL-7 plasma levels are generally observed in HIV-infected patients. IL-7 may also support bioenergetics that control cell fate decisions in lymphocytes. Recombinant Human IL-7 produced in E. coli is a single, non-glycosylated polypeptide chain containing 153 amino acids and having a molecular mass of 17,412 Daltons.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty