Would you like to log in again?
IL-17A Human Recombinant Protein, Rockland Immunochemicals
Supplier: Rockland Immunochemical
009-001-B32
RL009-001-B32EA 311.76 USD InStock
RL009-001-B32
IL-17A Human Recombinant Protein, Rockland Immunochemicals
Proteins and Peptides
Interleukin17-A is a proinflammatory cytokine produced by activated T cells and exists as a homodimer. This cytokine regulates the activities of NF-kappaB and mitogen-activated protein kinases. IL-17A can stimulate the expression of IL6 and cyclooxygenase-2 (PTGS2/COX-2), as well as enhance the production of nitric oxide (NO). High levels of this cytokine are associated with several chronic inflammatory diseases including rheumatoid arthritis, psoriasis and multiple sclerosis. IL17-A is the founding member of a group of cytokines called the IL-17 family. IL17-A was originally identified as a transcript from a rodent T-cell hybridoma. To elicit its functions, IL17 binds to a type I cell surface receptor called IL17R of which there are at least three variants: IL17RA, IL17RB, and IL17RC.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty