Would you like to log in again?
IL-17F Human Recombinant Protein, Rockland Immunochemicals
Supplier: Rockland Immunochemical
009-001-B31
RL009-001-B31EA 311.76 USD InStock
RL009-001-B31
IL-17F Human Recombinant Protein, Rockland Immunochemicals
Proteins and Peptides
Recombinant Human Interleukin-17-F is a non-glycosylated protein consisting of two 134 amino acid polypeptide chains that are cysteine linked homodimeric polypeptide chains with a molecular weight of 30 kDa. Homodimeric interleukin-17F is a member of the IL-17 family of six homodimeric proteins (IL-17A-17F). Activity studies reveal IL-17A to be the most potent, followed by IL-17A/F and IL-17F 100 fold lower than IL-17A.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty