Would you like to log in again?
IL-6 Human Recombinant Protein, Rockland Immunochemicals
Supplier: Rockland Immunochemical
009-001-310
RL009-001-310EA 293.94 USD InStock
RL009-001-310
IL-6 Human Recombinant Protein, Rockland Immunochemicals
Proteins and Peptides
Cytokines of the IL6/GCSF/MGF family are glycoproteins of about 170 to 180 amino acid residues that contains four conserved cysteine residues involved in two disulfide bonds. They have a compact, globular fold (similar to other interleukins), stabilized by the 2 disulfide bonds. One half of the structure is dominated by a 4 alpha-helix bundle with a left-handed twist: the helices are anti-parallel, with 2 overhand connections, which fall into a 2-stranded anti-parallel beta-sheet. The fourth alpha-helix is important to the biological activity of the molecule. Interleukin (IL)-6 is an important proinflammatory and immunoregulatory cytokine expressed by various cells. Interleukin-6 has been shown to inhibit the growth of early stage and to promote the proliferation of advanced stage melanoma cells in vitro. Recombinant Human IL-6 produced in E.coli is a single, non-glycosylated polypeptide chain containing 184 amino acids and having a molecular mass of 21,000 Daltons.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty