Would you like to log in again?
CyclinD1 protein-GST fusion, Rockland Immunochemicals
Supplier: Rockland Immunochemical
009-001-153 009-001-153S
10801-062 10801-060
CyclinD1 protein-GST fusion, Rockland Immunochemicals
CyclinD1 is a member of the cyclin family of proteins that are important regulators cyclin-dependent protein kinases (CDKs) (1). CyclinD1 forms a complex and functions as a regulatory subunit of CDK4 or CDK6 which are important during cell cycle G1/S transition. CyclinD1 has been shown to interact with the tumor suppressor protein Rb and the expression of CyclinD1 is positively regulated by Rb. Mutations, amplification and overexpression of CyclinD1 have been frequently observed in a variety of tumors and suggesting a role of this protein in tumorigenesis (2). CyclinD1 is ideal for investigators involved in Signaling Proteins, Cell Cycle Proteins, Cancer, and Cell Cycle research.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty