Would you like to log in again?
DbpA Control Protein, Rockland Immunochemicals
Supplier: Rockland Immunochemical
000-001-B98
RL000-001-B98EA 414.19 USD InStock
RL000-001-B98
DbpA Control Protein, Rockland Immunochemicals
Proteins and Peptides
DbpA, or Decorin Binding Protein A is from the spirochete Borrelia burgdorferi, which is carried by Ixodes ticks. DbpA from other microbial organisms such as E. coli (ATP-dependent RNA helicase DbpA) are significantly different. The spirochete migrates from the tick midgut during tick feeding to tick salivary glands and are thus transmitted to the mammal host. This transition may be facilitated by changes in expression of some B. burgdorferi genes. Spirochetal surface adhesions mediate attachment to decorin, a major component of the host extracellular matrix, enabling bacteria to colonize in mammalian tissues. It is believed that expression of the various proteins associated with the spirochete may be regulated by the changes in tick life cycle, changes in conditions during tick feeding (such as temperature, pH, and nutrients) and/or in coordination with the course of infection of the mammal host. Lyme disease proteins are ideal for researchers interested in immunology, neurology, and rheumatology, coinfections , autoimmune, and neurodegenerative diseases.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty