Would you like to log in again?
TSG Series Chromatography Refrigerators, Thermo Scientific
Supplier: Thermo Scientific
TSG30CPGA TSG72CSGA TSG45CSLA TSG30CSGA TSG72CPGA TSG49CSGA TSG45CPLA TSG49CPGA
76299-480EA 8022.73 USD InStock
76299-480 76299-494 76299-486 76299-482 76299-492 76299-490 76299-484 76299-488
TSG Series Chromatography Refrigerators, Thermo Scientific
Refrigerators
TSG Series Chromatography Refrigerators are designed with chromatography applications in mind. These refrigerators are designed with the performance, security, and quality features for everyday laboratory needs.
  • Featuring a wide range of cabinet sizes from 30 cu. ft. to 72 cu. ft.
  • Hydrocarbon safety sensor protected interior duplex outlet
  • Whisper-quiet operation at 51 dBA
  • Green, natural refrigerants
  • Choice of white painted interior and exterior or stainless-steel interior and gray exterior
  • Digital controller with Temperature, door ajar, and power failure (with battery back-up) alarms
  • Two years parts and labor, plus three years compressor warranty
These refrigerators are features with rear access port, plus two 2-inch side access ports, keyed door lock(s), remote alarm contacts, LED interior lighting, double pane glass doors.
These are ideal for the demanding requirements of daily use in research, clinical, and industrial laboratories storing reagents, media, proteins, and samples used daily in the lab.
Certifications: cULus, EPA ENERGY STAR® certified.
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty