Would you like to log in again?
TexTab™ Disinfectant, ITW Texwipe
Supplier: ITW-TEXWIPE COMPANY
TX6460 TX6460
75982-220CS 520.87 USD InStock
CA75982-220 75982-220
TexTab™ Disinfectant, ITW Texwipe
Cleaning Agents Disinfectants
TexTab™ is a bleach alternative in an effervescent tablet form.
  • Disinfecting any hard, non-porous, inanimate surface
  • Replacing liquid bleach solutions in all applications
  • Pre-cleaning gross soil before disinfection
  • Cleaning and disinfecting small work areas as well as large areas (floors, walls, ceilings)
  • Fogging
Using as part of a disinfectant rotation program as a bactericidal disinfectant for everyday use and/or a sporicidal disinfectant weekly or biweekly. Dilution chart can be used for different dilution rates.
Kills C. difficile (C. diff) spores in just 4 minutes.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty