Would you like to log in again?
Rotavapor® R-250, Buchi
Supplier: BUCHI CORPORATION
8501D552 8500G542
76266-282EA 146857 USD InStock
76266-282 76266-280
Rotavapor® R-250, Buchi
Evaporators Rotary Evaporators
Offering the highest performance, efficiency and safety for industrial distillation, the Rotavapor® R-250 is a long lasting investment.
  • Electric heating bath for water or oil, temperature range 20 to 180 °C
  • Display of bath temperature, vapor temperature and rotation speed
  • Easy connection of evaporating flask with patented snap flange coupling
  • Complete inert materials with borosilicate glass and PTFE
The Rotavapor® R-250 offers fast distillation of large batches in flasks up to 50 L. A wide range of accessories and glass configurations meets the needs of every application. The system is easy to use thanks to intuitive operation of the Rotavapor® and it's large, easy to read vacuum controller display. Expect a long life from your investment due to exclusive use of corrosion-free stainless steel, glass and PTFE components.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty