Would you like to log in again?
Reveleris® X2 UV Flash Chromatography System, Buchi
Supplier: BUCHI CORPORATION
140000014
76266-272EA 34661.32 USD InStock
76266-272
Reveleris® X2 UV Flash Chromatography System, Buchi
Chromatography Systems Flash Chromatography Systems
The Reveleris® X2-UV flash chromatography system enables fast and easy purification covering the essential needs of organic chemists regardless of complexity of the sample.
  • Quickly modify run parameters on a large 12" touch screen display
  • Edit gradients in real time, using click and drag or table
  • Easily switch between normal phase and reversed-phase applications
  • Avoid running out of solvent with included solvent level sensors
  • Auto-recognition of cartridges and trays reduces set-up time, and the risk of potential errors
The Reveleris® X2-UV flash chromatography system allows for ultimate flexibility in applications with four independent solvent channels for binary gradients (linear, step and isocratic). Method parameters are suggested by cartridge size and type with RfID technology or by manual input. Three UV wavelengths can be used to collect fractions in tubes with auto rack recognition.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty