Would you like to log in again?
Vydac® 150HC C18 HPLC Column, Hichrom
Supplier: HICHROM LIMITED
Vydac®
5168320
75799-114EA 622.41 USD InStock
75799-114
Vydac® 150HC C18 HPLC Column, Hichrom
Chromatography Columns
Vydac® 150HC column is media, developed specifically for peptide purification.
  • Robust, high capacity 150 Å silica bonded with C18 for reversed phase
This is highly effective for purifying many small to medium-sized peptides with greater loading capacity and improved productivity.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty