Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Propanol ≥99.8% (by GC) USP, NF, ACS

Supplier: Pharmco-Aaper TE

Danger
Synonyms: Isopropyl alcoholIsopropyl Rubbing Alcoholsec-Propanol2-Hydroxy propanesec-Propyl AlcoholDimethyl carbinol2-HydroxypropanePropane, 2-hydroxy-Propan-2-olIPAIsopropanol

231USP/NFCSGL 231000099DM55
76239-624 76242-616
2-Propanol ≥99.8% (by GC) USP, NF, ACS


Formula: (CH₃)₂CHOH
MW: 60,1 g/mol
Boiling Pt: 82 °C (1013 hPa)
Melting Pt: –89 °C
Density: 0.786 g/cm³ (20 °C)
Flash Pt: 12 °C
Storage Temperature: ambient
MDL Number: MFCD00011674
CAS Number: 67-63-0
EINECS: 200-661-7
UN: 1219
ADR: 3,II
REACH: 01-2119457558-25
Merck Index: 13,05228
https://us.vwr.com/stibo/web/std.lang.all/19/69/731969.jpg
Order Now
Specification Test Results
ACS/USP - Assay by GC Min. 99.8%
ACS/USP - Assay by GC Corrected for Water Min. 99.8%
USP - Identification (IR) Passes Test
ACS - Color (APHA) NMT 10
ACS - Residue After Evaporation 10 ppm
USP - Limit of Non-Volatile Residue NMT 2.5 mg
ACS - Titrable Acid and Base Max. 0.0001 meq/g
USP - Acidity NMT 0.70 ml of 0.02 N NaOH Required for Neutralization
ACS - Water 0.2%
ACS - Carbonyl Compounds 0.002% Max. Each of Propionaldehyde or Acetone 0.002%
USP - Refractive Index at 20 °C 1.376 - 1.378
USP - Specific Gravity at 25 °C 0.783 - 0.787


 

ORDER

Q and A

Product Questions and Answers

Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty