Would you like to log in again?
Due to system maintenance, custom pricing and product availability will not be shown from 7:00pm EST Friday through 6:00pm Sunday. Orders placed will be processed after maintenance is complete.
Vented Duocap® for Flasks, Triforest Labware
Supplier: TRIFOREST ENTERPRISES INC
DCPC-38S DCPC-53S
10014-828CS 791.27 USD InStock
10014-828 10014-830
Vented Duocap® for Flasks, Triforest Labware
Closures Bottle Closures
Vented Duocap® is constructed of standard blue polypropelene with a PTFE membrane that is ideal for aerobic/anaerobic cell culture.
  • Autoclavable
  • Two caps in one
  • Dustguard cover allows sterile exchange
  • Comes standard on both flat and baffled flasks of all sizes
There is no hassle or risk associated with switching caps.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty