Would you like to log in again?
LB Agar Plates with 150 µg/ml Ampicillin, Teknova
Supplier: TEKNOVA INC
L6005 L6005
101320-154PK 106.54 USD InStock
101320-154 CA101320-154
LB Agar Plates with 150 µg/ml Ampicillin, Teknova
Media for Microbiology Prepared Media for Microbiology
For use with recombinant strains of E. coli containing plasmids with the resistance gene to ampicillin (bla or ampR) such as pBluescript, pGEM, or the pUC series of plasmids.
  • Shipping at room temperature
  • Store at 2 to 8 °C
Contains 1.0% tryptone (10.0 g/l); 0.5% yeast extract (5.0 g/l); 1.0% sodium chloride (NaCl) (10.0 g/l); 1.5% agar (15.0 g/l) and 150 µg/ml ampicillin.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty