LB Broth with Chloramphenicol, Teknova

Supplier: Teknova
L8130 L8130
100219-750EA 82.57 USD
CA100219-750 100219-750
LB Broth with Chloramphenicol, Teknova
Media for Microbiology Dehydrated Media for Microbiology
LB broth with chloramphenicol is designed for the maintenance and propagation of chloramphenicol resistant strains and strains harboring plasmids such as pBC Ks +/-, pGV1124, and pJAK16 series.

  • Shipping on ice
  • Store at frozen condition

Contains 1.0% tryptone, 0.5% yeast extract, 1.0% NaCl with 50 µg/ml chloramphenicol.
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR