Would you like to log in again?
Vogel Johnson Agar Plates, Teknova
Supplier: TEKNOVA INC
V0144 V0144
101320-726PK 110.62 USD InStock
101320-726 CA101320-726
Vogel Johnson Agar Plates, Teknova
Media for Microbiology Prepared Media for Microbiology
For the selective isolation of coagulase-positive, mannitol-positive Staphylococcus aureus.
  • Shipping at ambient
  • Store at room temperature
Contains 1.0% trytone, 0.5% yeast extract, 1.0% mennitol, 0.5% dye potassium phosphate, 0.5% lithium chloride., 1.0% glycene, 1.5% agar and 0.0025% phenol red.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty