Would you like to log in again?
Anti-Muscarinic Cholinergic Receptor 4 (M4) Antibody ELISA Assay Kit, Eagle Biosciences
Supplier: Eagle Biosciences
M4A31-K01
103573-006EA 2105.38 USD InStock
103573-006
Anti-Muscarinic Cholinergic Receptor 4 (M4) Antibody ELISA Assay Kit, Eagle Biosciences
Assays ELISAs
The Eagle Bio Anti-Muscarinic Cholinergic Receptor 4 (M4) Antibody ELISA is for research use only.
  • Sensitivity: Cut-off
  • Dynamic Range: 2.5 to 40 U/ml
  • Sample Size: 100 μl
  • Sample Type: Serum
  • Incubation Time: 3.5 hours
Muscarinic cholinergic receptors, or mAChRs, areacetylcholine receptors that form G protein-receptorcomplexes in the cell membranes of certain neuronsand other cells. Autoantibodies against M4 AChR aresignificantly elevated in patients suffering fromchronic fatigue syndrome (CFS) / myalgicencephalomyelitis (ME). Muscarinic cholinergicreceptor 4 (M4) autoantibodies are present in asubset of 20-30% of chronic fatigue syndrome patients.
The Eagle Biosciences anti-muscarinic cholinergic receptor 4(M4)-Antibody EIA is designed for the determination of antibodies against the muscarinic cholinergic receptor 4 (M4) in serum.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty