Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-Muscarinic Cholinergic Receptor 3 (M3) Antibody ELISA Assay Kit, Eagle Biosciences

Supplier: Eagle Biosciences

M3A31-K01
103573-004EA 2105.38 USD InStock
103573-004
Anti-Muscarinic Cholinergic Receptor 3 (M3) Antibody ELISA Assay Kit, Eagle Biosciences
Assays ELISAs
The Eagle Bio Anti-Muscarinic Cholinergic Receptor 3 (M3) Antibody ELISA is for research use only.

  • Sensitivity: Cut-off
  • Dynamic Range: 2.5 to 40 U/ml
  • Sample Size: 100 μl
  • Sample Type: Serum
  • Incubation Time: 3.5 hours

Muscarinic cholinergic receptors, or mAChRs, are acetylcholine receptors that form G protein-receptor complexes in the cell membranes of certain neurons and other cells. Autoantibodies against M3 AChR are significantly elevated in patients suffering from chronic fatigue syndrome (CFS) / myalgic encephalomyelitis (ME). Muscarinic acetylcholine receptor 3 (M3) autoantibodies are present in a subset of 20 to 30% of chronic fatigue syndrome patients.

The Eagle Biosciences anti-muscarinic cholinergic receptor 3 (M3)-antibody EIA is designed for the determination of antibodies against the muscarinic cholinergic receptor 3 (M3) in serum.

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty