Would you like to log in again?
NT-proCNP ELISA, Eagle Biosciences
Supplier: Eagle Biosciences
BI-20812
103573-002EA 784.62 USD InStock
103573-002
NT-proCNP ELISA, Eagle Biosciences
Assays ELISAs
The NT-proCNP ELISA is intended for quantitative determination of human NT proCNP in plasma, serum.
  • Sensitivity: 0.7 pmol/l
  • Dynamic Range: 4 to 128 pmol/l
  • Sample Size: 20 µl
  • Sample Type: Serum, EDTA plasma, heparin plasma, and citrate plasma
  • Incubation Time: 4 hours
CNP belongs to a well characterised group of natriuretic peptides like atrial natriuretic (ANP) and brain natriuretic peptide (BNP), which play various important roles as regulatory mechanisms as well as bio-markers. They participate in the regulation of blood pressure and body fluid homeostasis, modify growth and development of cardiovascular tissues and bone. ANP and BNP are cardiac hormones, secreted by the atria and ventricles, respectively, in the normal adult heart.
In contrast to ANP and BNP, very little CNP is produced by cardiac tissue, but is expressed primarily in the brain, pituitary gland, vascular endothelium, kidney and female reproductive tract.All natriuretic peptides are produced as propeptides, which are subsequently cleaved into the biologically active, C-terminal hormone and the N-terminal fragment (NT-proANP 1-98, NT-proBNP 1-76 and NT-proCNP, which length is still under discussion). Since those N-terminal fragments are much more stable, and circulate in higher amounts than the active hormones, they are easier and more reliable to be measured in serum or plasma. So we developed a sandwich immunoassay for NT-proCNP using antibodies directed against amino acids 1-19 and 30-50.
The physiological role of CNP is not only studied in cardiac disease, but also in bone developmental biology, generally in bone research, renal diseases, embryonic developmental research and vascular diseases.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty