Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Zinc chloride 98.0-100.5% (by complexometric analysis), EMPROVE® API Ph. Eur., BP, USP

Supplier: MilliporeSigma

Danger
Synonyms: Zinc (II) chlorideButter of zinc

1.08815.1000 1.08815.1000
EM1.08815.1000CS 5232.35 USD InStock
EM1.08815.1000 CA1.08815.1000
Zinc chloride 98.0-100.5% (by complexometric analysis), EMPROVE® API Ph. Eur., BP, USP
Zinc chloride
Elemental impurity specifications have been set considering ICH Q3D (Guideline for Elemental Impurities).
Class 1-3 elements are not likely to be present above the ICH Q3D option 1 limit, unless specified and indicated (*).

Conforms to Ph Eur, BP, USP


Formula: ZnCl₂
MW: 136,3 g/mol
Boiling Pt: 732 °C (1013 hPa)
Melting Pt: 287 - 304 °C
Density: 2.93 g/cm³ (22 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011295
CAS Number: 7646-85-7
EINECS: 231-592-0
UN: 2331
ADR: 8,III
Merck Index: 12,10261
Order Now
Specification Test Results
Assay (Complexometric, ZnCl₂) 98.0 - 100.5%
Identity (Zn) Passes Test
Identity (Cl) Passes Test
pH-value (10% water) 4.6 - 5.5
Oxychloride Passes Test
Sulfate (SO₄) ≤0.01%
Total Nitrogen (N) ≤0.001%
Aluminium, Calcium, Magnesium, Heavy metals, Iron Passes Test
As (Arsenic) ≤0.0002%
Ca (Calcium) ≤0.01%
Cd (Cadmium) ≤0.0005%
Fe (Iron) ≤0.001%
K (Potassium) ≤0.15%
Na (Sodium) ≤0.01%
NH₄ (Ammonium) ≤0.04%
Pb (Lead) ≤0.005%
Tl (Thallium) ≤0.0005%
Alkali- and Alkaline Earth Metals (as SO₄) ≤1.0%
Residual Solvents (Ph. Eur./USP/ICH) Excluded by Manufacturing Process


 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty