Sodium acetate, anhydrous 99.0-101.0% (dry basis), EMPROVE® EXPERT USP for biopharmaceutical production, SAFC®

Synonyms: Acetic acid sodium salt

1.37046.5000 1.37046.5000 1.37046.9012 1.37046.1000 1.37046.9012 1.37046.9100
EM1.37046.5000EA 530.12 USD
CA1.37046.9012 EM1.37046.5000 CA1.37046.5000 EM1.37046.1000 EM1.37046.9012 EM1.37046.9100
Sodium acetate, anhydrous 99.0-101.0% (dry basis), EMPROVE® EXPERT USP for biopharmaceutical production, SAFC®
Sodium acetate

Elemental impurity specifications have been set considering ICH Q3D (Guideline for Elemental Impurities). Class 1-3 elements are not likely to be present above the ICH Q3D option 1 limit, unless specified and indicated (*).


Corresponds to USP

Formula: H₃CCOONa
MW: 82.03 g/mol
Boiling Pt: 881.4 °C (1013 hPa)
Melting Pt: 324 °C
Density: 1.52 g/cm³ (20 °C)
Flash Pt: >250 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00012459
CAS Number: 127-09-3
EINECS: 204-823-8

Order Now

Specification Test Results

Assay (Perchloric Acid Titration, Calculated on Dried Substance) 99.0 - 101.0%
Identity Passes Test
In Water Insoluble Matter ≤0.05%
pH-Value (3%, Water) 7.5 - 9.2
Chloride (Cl) ≤0.01%
Sulfate (SO₄) ≤0.005%
Heavy Metals (as Pb) ≤0.001%
Al (Aluminium) ≤0.00002%
K (Potassium) ≤0.003%
Calcium and Magnesium Passes Test
Assay (Acetic Acid) ≤0.4%
Loss on Drying (120 °C) ≤1.0%
Other Residual Solvents (USP/ICH) Excluded by Manufacturing Process
Endotoxins ≤2.5 I.U./g
Total Aerobic Microbial Count ≤100 CFU/g
Total Yeast and Mould Count ≤100 CFU/g

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR