Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

EDTA (ethylenediamine tetraacetic acid) 98.0-100.5% (by complexometric analysis) Ph. Eur., NF

Supplier: MilliporeSigma

Warning
Synonyms: Ethylenediaminetetraacetic acidIdranal® IIKestranal® AEdetic AcidEDTAEdetic acidEthylenedinitrilotetra-acetic acid

1.00944.9025
EM1.00944.9025EA 1593.47 USD InStock
EM1.00944.9025
EDTA (ethylenediamine tetraacetic acid) 98.0-100.5% (by complexometric analysis) Ph. Eur., NF
EDTA (Ethylenediamine tetraacetic acid)


Formula: C₁₀H₁₆N₂O₈
MW: 292,25 g/mol
Boiling Pt: 614 °C (1013 hPa)
Melting Pt: 220 °C
Density: 0.86 g/cm³ (20 °C)
Flash Pt: Min. 100 °C
Storage Temperature: ambient
MDL Number: MFCD00003541
CAS Number: 60-00-4
EINECS: 200-449-4
UN: 0000
REACH: 01-2119486399-18
Merck Index: 13,03546
https://us.vwr.com/stibo/web/std.lang.all/72/41/2297241.jpg
Order Now
Specification Test Results
Assay (Complexometric) 98.0 - 100.5%
Identity (IR Spectrum) Conforms
Chloride (Cl) ≤0.02%
Heavy Metals (as Pb) ≤0.002%
Fe (Iron) ≤0.005%
Nitrilotriacetic Acid (HPLC) ≤0.1%
Formaldehyde ≤0.001%
Other Residual Solvents (Ph. Eur./USP/ICH) Excluded by Production Process
Sulfated Ash (600 °C) ≤0.2%


 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty