Would you like to log in again?
LB Agar Plates with Animal-Free Soytone and Ampicillin 100, Teknova
Supplier: TEKNOVA INC
L1094 L1094
100216-948PK 107.92 USD InStock
CA100216-948 100216-948
LB Agar Plates with Animal-Free Soytone and Ampicillin 100, Teknova
Media for Microbiology Prepared Media for Microbiology
For use with recombinant E. coli strains harboring plasmids such as pBluescript, pGEM, pUC series plasmids. Used when the media cannot contain any animal products.
Contains 0.5% yeast extract, 1.0% sodium chloride, 1.5% agar, 100 µg/ml ampicillin, 1.69% animal-free soytone.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty