Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-ITGAV & ITGB6 Heterodimer Mouse Monoclonal Antibody [clone: 06]

Supplier: Sino Biological

CT039-MM06-200 CT039-MM06-100 CT039-MM06-50
103662-554EA 214.31 USD InStock
103662-554 103662-216 103589-940
Anti-ITGAV & ITGB6 Heterodimer Mouse Monoclonal Antibody [clone: 06]
Antibodies
Anti-CD51 & ITGB6 Heterodimer

Type: Primary
Antigen: ITGAV & ITGB6 Heterodimer
Clonality: Monoclonal
Clone: Clone: 06
Conjugation:
Epitope:
Host: Mouse
Isotype: Mouse IgG1
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty