Anti-IL-23 Mouse Monoclonal Antibody [clone: 06]

Supplier: Sino Biological

CT037-MM06-50 CT037-MM06-100 CT037-MM06-200
103589-938EA 106.62 USD
103589-938 103662-212 103662-214
Anti-IL-23 Mouse Monoclonal Antibody [clone: 06]
Antibodies
Anti-IL-23 (IL12B & IL23A Hete

Type: Primary
Antigen: IL-23
Clonality: Monoclonal
Clone: Clone: 06
Conjugation:
Epitope:
Host: Mouse
Isotype: Mouse IgG2a
Reactivity: Marmoset
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR