Anti-FCGRT & B2M Heterodimer A Rabbit Monoclonal Antibody [clone: 101]

Supplier: Sino Biological
CT030-R101-50 CT030-R101-100
103589-932EA 119 USD
103589-932 103662-644
Anti-FCGRT & B2M Heterodimer A Rabbit Monoclonal Antibody [clone: 101]
Antibodies
Anti-FCGRT & B2M Heterodimer A

Type: Primary
Antigen: FCGRT & B2M Heterodimer A
Clonality: Monoclonal
Clone: Clone: 101
Conjugation:
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR