Anti-IL-23 Rabbit Polyclonal Antibody

Supplier: Sino Biological

CT028-RP01-100 CT028-RP01-200 CT028-RP01-400
103662-208EA 106.62 USD
103662-208 103662-210 103662-642
Anti-IL-23 Rabbit Polyclonal Antibody
Antibodies
Anti-IL-23 (IL12B & IL23A Hete

Type: Primary
Antigen: IL-23
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR