Anti-CD3D/CD3E Rabbit Monoclonal Antibody (APC (Allophycocyanin)) [clone: 301]

Supplier: Sino Biological

CT026-R301-A-25 CT026-R301-A-100
103630-284EA 26.92 USD
103630-284 103632-480
Anti-CD3D/CD3E Rabbit Monoclonal Antibody (APC (Allophycocyanin)) [clone: 301]
Antibodies
Anti-CD3D & CD3E Heterodimer A

Type: Primary
Antigen: CD3D/CD3E
Clonality: Monoclonal
Clone: Clone: 301
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR