Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-ITGA5 & CD29 Heterodimer Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: 05]

Supplier: Sino Biological

CT014-MM05-F-100 CT014-MM05-F-25
103632-476EA 150.77 USD InStock
103632-476 103630-280
Anti-ITGA5 & CD29 Heterodimer Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: 05]
Antibodies
Anti-ITGA5 & CD29 Heterodimer

Type: Primary
Antigen: ITGA5 & CD29 Heterodimer
Clonality: Monoclonal
Clone: Clone: 05
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Mouse
Isotype: Mouse IgG2b
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty