Anti-ITGA5 and ITGB1 Mouse Monoclonal Antibody [clone: 05] APC (Allophycocyanin)

Supplier: Sino Biological
CT014-MM05-A-25 CT014-MM05-A-100
103630-278EA 90.9 USD
103630-278 103632-474
Anti-ITGA5 and ITGB1 Mouse Monoclonal Antibody [clone: 05] APC (Allophycocyanin)
Antibodies
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human ITGA5 and ITGB1 Heterodimer (rh ITGA5 and ITGB1 Heterodimer; Catalog#CT014-H2508H; P08648 and P05556-1; Met1-Tyr995 and Met1-Asp728) and conjugated with APC under optimum conditions, the unreacted APC was removed.

Type: Primary
Antigen: ITGA5 & CD29 Heterodimer
Clonality: Monoclonal
Clone: Clone: 05
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: Mouse IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR