Anti-ITGA6 and ITGB1 Rabbit Polyclonal Antibody

Supplier: Sino Biological
CT013-RP01-400 CT013-RP01-200 CT013-RP01-100
103662-170EA 562.97 USD
103662-170 103662-552 103662-550
Anti-ITGA6 and ITGB1 Rabbit Polyclonal Antibody
Antibodies
Produced in rabbits immunized with purified, recombinant Human ITGA6 and ITGB1 Heterodimer (rh ITGA6 and ITGB1 Heterodimer; Catalog#CT013-H2508H; P23229-3 and P05556-1; Met 1-Ser 1011, Met 1-Asp 728). Total IgG was purified by Protein A affinity chromatography.

Type: Primary
Antigen: ITGA6 & ITGB1 Heterodimer
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR