Anti-IL12B and IL23A Rabbit Polyclonal Antibody

Supplier: Sino Biological
CT012-RP01-200 CT012-RP01-100 CT012-RP01-400
103662-546EA 562.97 USD
103662-546 103662-544 103662-542
Anti-IL12B and IL23A Rabbit Polyclonal Antibody
Antibodies
Produced in rabbits immunized with purified, recombinant Human IL-23 (IL23A and IL12B Heterodimer) (Catalog#CT012-H08H;NP_057668.1 and NP_002178.2; Met1-Pro189 and Met1-Ser328). Total IgG was purified by Protein A affinity chromatography.

Type: Primary
Antigen: IL-23
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR