Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-FCGRT & B2M Heterodimer Rabbit Polyclonal Antibody

Supplier: Sino Biological

CT009-T08-50 CT009-T08-200 CT009-T08-100
103637-996EA 106.62 USD InStock
103637-996 103589-890 103589-888
Anti-FCGRT & B2M Heterodimer Rabbit Polyclonal Antibody
Antibodies
Anti-FCGRT & B2M Heterodimer A

Type: Primary
Antigen: FCGRT & B2M Heterodimer
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty