Anti-VISTA Rabbit Polyclonal Antibody

Supplier: Sino Biological

90801-RP01-200 90801-RP01-100 90801-RP01-400
103662-148EA 160.46 USD
103662-148 103662-150 103662-152
Anti-VISTA Rabbit Polyclonal Antibody
Antibodies
Anti-VISTA Antibody

Type: Primary
Antigen: VISTA
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Rhesus Monkey
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR