Anti-TNFA Rabbit Polyclonal Antibody

Supplier: Sino Biological

90330-RP01-200 90330-RP01-100 90330-RP01-400
103662-138EA 160.46 USD
103662-138 103662-136 103662-140
Anti-TNFA Rabbit Polyclonal Antibody
Antibodies
Anti-TNF-alpha Antibody

Type: Primary
Antigen: TNFA
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Cynomolgus
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR