Anti-EGFR Rabbit Polyclonal Antibody

Supplier: Sino Biological
90317-T16-200 90317-T16-100 90317-T16-50
103589-882EA 270.87 USD
103589-882 103589-884 103637-994
Anti-EGFR Rabbit Polyclonal Antibody
Antibodies
Produced in rabbits immunized with purified, recombinant Rhesus EGFR/HER1/ErbB1 ( Catalog#90317-K08H; XP_001107305.1; Met1-Ser645). EGFR/HER1/ErbB1 specific IgG was purified by Rhesus EGFR/HER1/ErbB1 affinity chromatography.

Type: Primary
Antigen: EGFR
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Rhesus Monkey
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR