Anti-IL21R Rabbit Polyclonal Antibody

Supplier: Sino Biological
90315-RP01-100 90315-RP01-200 90315-RP01-400
103662-536EA 562.97 USD
103662-536 103662-534 103662-532
Anti-IL21R Rabbit Polyclonal Antibody
Antibodies
Produced in rabbits immunized with purified, recombinant Cynomolgus IL-21R/Il21R (Catalog#90315-C08H; XP_005591578.1; Met1-Pro236). Total IgG was purified by Protein A affinity chromatography.

Type: Primary
Antigen: IL-21R
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Cynomolgus
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR