Anti-TGFB1 Rabbit Polyclonal Antibody

Supplier: Sino Biological

90294-T16-50 90294-T16-200 90294-T16-100
103637-982EA 106.62 USD
103637-982 103589-860 103589-858
Anti-TGFB1 Rabbit Polyclonal Antibody
Antibodies
Anti-TGF beta 1 Antibody

Type: Primary
Antigen: TGFB1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Cynomolgus
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR