Anti-IL-8 Rabbit Polyclonal Antibody

Supplier: Sino Biological

90280-RP01-400 90280-RP01-200 90280-RP01-100
103662-112EA 214.31 USD
103662-112 103662-110 103662-108
Anti-IL-8 Rabbit Polyclonal Antibody
Antibodies
Anti-IL-8 Antibody

Type: Primary
Antigen: IL-8
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Rhesus Monkey
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR