Anti-IL-8 Rabbit Monoclonal Antibody [clone: 009]

Supplier: Sino Biological

90280-R009-100 90280-R009-50
103662-106EA 214.31 USD
103662-106 103592-252
Anti-IL-8 Rabbit Monoclonal Antibody [clone: 009]
Antibodies
Anti-IL-8 Antibody

Type: Primary
Antigen: IL-8
Clonality: Monoclonal
Clone: Clone: 009
Conjugation:
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Rhesus Monkey
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR